Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluorobenzoyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1863T-100G PC1863T-25G PC1863T-5G
77413-360EA 124.3 USD InStock
77413-360 77413-362 77413-364
2-Chloro-4-fluorobenzoyl chloride 98%
2-Chloro-4-fluorobenzoyl chloride


Formula: C₇H₃Cl₂FO
MW: 193 g/mol
Boiling Pt: 82…84 °C/15 mmHg
Density: 1.447 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00084954
CAS Number: 21900-54-9
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty