Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluorobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2615A-100G PC2615A-10G PC2615A-25G
77413-728EA 167.08 USD InStock
77413-728 77413-730 77413-732
2,3-Difluorobenzaldehyde 98%
2,3-Difluorobenzaldehyde


Formula: C₇H₄F₂O
Boiling Pt: 70 °C/15 mmHg
Density: 1.301 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00010292
CAS Number: 2646-91-5
UN: 1989
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty