Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-iodobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC3784-100G PC3784-10G PC3784-25G
77414-884EA 119.2 USD InStock
77414-884 77414-886 77414-888
2-Fluoro-6-iodobenzonitrile 98%
2-Fluoro-6-iodobenzonitrile


Formula: C₇H₃FIN
MW: 247.01
Boiling Pt: 281
Melting Pt: 48…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00015478
CAS Number: 79544-29-9
UN: 3439
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty