Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,2-Dibromo-5-fluorotoluene 98+%

Supplier: APOLLO SCIENTIFIC
Danger

PC4075-25G PC4075-5G
77415-082EA 27.5 USD InStock
77415-082 77415-084
α,2-Dibromo-5-fluorotoluene 98+%
α,2-Dibromo-5-fluorotoluene


Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Boiling Pt: 89…90 °C/1 mmHg
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00042187
CAS Number: 112399-50-5
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty