Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3,4,4-Hexafluorobutyric acid

Supplier: APOLLO SCIENTIFIC
Danger

PC408040-250MG
77415-092EA 21.4 USD InStock
77415-092
2,2,3,3,4,4-Hexafluorobutyric acid
2,2,3,3,4,4-Hexafluorobutyric acid


Formula: C₄H₂F₆O₂
Boiling Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00465636
CAS Number: 679-12-9
UN: 1760
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty