Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Difluoro-1-nitro-2-propanol 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC450554-1G PC450554-250MG
77415-556EA 1118.66 USD InStock
77415-556 77415-558
3,3-Difluoro-1-nitro-2-propanol 95%
3,3-Difluoro-1-nitro-2-propanol


Formula: C₃H₅F₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD28137318
CAS Number: 148628-50-6
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty