Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-(trifluoromethoxy)benzylamine 98%

Supplier: APOLLO SCIENTIFIC

PC302759-5G PC302759-1G
77414-256EA 1272.5 USD InStock
77414-256 77414-252
5-Chloro-2-(trifluoromethoxy)benzylamine 98%
5-Chloro-2-(trifluoromethoxy)benzylamine


Formula: C₈H₇ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD11519365
CAS Number: 874821-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty