Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S)-(-)-3,3,3-Trifluoro-1,2-propenoxide 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC3905-1G PC3905-250MG
77415-008EA 289.34 USD InStock
77415-008 77415-010
(2S)-(-)-3,3,3-Trifluoro-1,2-propenoxide 97%
(2S)-(-)-3,3,3-Trifluoro-1,2-propenoxide


Formula: C₃H₃F₃O
MW: 112.051 g/mol
Boiling Pt: 25…32 °C
Melting Pt: –80 °C
Density: 1.294 g/cm³ (25 °C)
Flash Pt: –26 °C
Storage Temperature: Freezer
MDL Number: MFCD04128982
CAS Number: 130025-34-2
UN: 1993
ADR: 3 I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty