Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Bis(trifluoromethyl)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC3104T-1G PC3104T-25G PC3104T-5G
77414-474EA 38.72 USD InStock
77414-474 77414-476 77414-478
2,4-Bis(trifluoromethyl)benzyl bromide
2,4-Bis(trifluoromethyl)benzyl bromide


Formula: C₉H₅BrF₆
MW: 307.03
Boiling Pt: 112 °C/0.2 mmHg
Density: 1.637
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00010306
CAS Number: 140690-56-8
UN: 2920
ADR: 8+3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty