Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Undecafluorohexanoic acid

Supplier: APOLLO SCIENTIFIC
Danger

PC3454-10G PC3454-1G PC3454-5G
77414-738EA 39.74 USD InStock
77414-738 77414-740 77414-742
Undecafluorohexanoic acid
Undecafluorohexanoic acid


Formula: C₆HF₁₁O₂
MW: 314.05 g/mol
Boiling Pt: 157 °C
Density: 1.762 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00198040
CAS Number: 307-24-4
EINECS: 206-196-6
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty