Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC31422-1G PC31422-250MG
77414-516EA 203.76 USD InStock
77414-516 77414-518
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride 97%
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride


Formula: C₁₁H₈ClFN₂O
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD03659694
CAS Number: 423768-49-4
UN: 1759
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty