Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexafluoroglutaric anhydride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC4710-25G PC4710-5G
77415-676EA 245.2 USD InStock
77415-676 77415-678
Hexafluoroglutaric anhydride 98%
Hexafluoroglutaric anhydride


Formula: C₅F₆O₃
Boiling Pt: 72 °C
Density: 1.654 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006680
CAS Number: 376-68-1
EINECS: 206-811-8
UN: 3265
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty