Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(trifluoromethyl)benzyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC4733-1G PC4733-25G PC4733-5G
77415-684EA 22.42 USD InStock
77415-684 77415-686 77415-688
2-Bromo-4-(trifluoromethyl)benzyl chloride
2-Bromo-4-(trifluoromethyl)benzyl chloride


Formula: C₈H₅BrClF₃
MW: 273.478 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094249
CAS Number: 480438-96-8
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty