Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Bromomethyl)-5-fluorobenzenesulfonyl chloride 90%

Supplier: APOLLO SCIENTIFIC
Danger

PC500582-1G PC500582-250MG
77416-016EA 379 USD InStock
77416-016 77416-018
2-(Bromomethyl)-5-fluorobenzenesulfonyl chloride 90%
2-(Bromomethyl)-5-fluorobenzenesulfonyl chloride


Formula: C₇H₅BrClFO₂S
MW: 287.534 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28096593
CAS Number: 1820717-41-6
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty