Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-fluorophenylacetylene 96%

Supplier: APOLLO SCIENTIFIC
Warning

PC56836-1G PC56836-250MG PC56836-5G
77416-882EA 54.5 USD InStock
77416-882 77416-884 77416-886
2-Bromo-5-fluorophenylacetylene 96%
2-Bromo-5-fluorophenylacetylene


Formula: C₈H₄BrF
MW: 199.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29046530
CAS Number: 1693887-36-3
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty