Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(trifluoromethyl)phenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC4376O-25G PC4376O-5G
77415-362EA 36.68 USD InStock
77415-362 77415-364
2-Fluoro-5-(trifluoromethyl)phenol 98%
2-Fluoro-5-(trifluoromethyl)phenol


Formula: C₇H₄F₄O
Boiling Pt: 146 °C
Density: 1.436 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00061286
CAS Number: 141483-15-0
UN: 2920
ADR: 8+3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty