Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluoro-5-iodoaniline 97%

Supplier: APOLLO SCIENTIFIC

PC48771-1G PC48771-5G
77415-790EA 406.5 USD InStock
77415-790 77415-794
2-Chloro-4-fluoro-5-iodoaniline 97%
2-Chloro-4-fluoro-5-iodoaniline


Formula: C₆H₄ClFIN
Storage Temperature: Refrigerator
MDL Number: MFCD31629890
CAS Number: 2091569-03-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty