Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-methoxybenzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC3817F-100G PC3817F-25G
77414-908EA 103.92 USD InStock
77414-908 77414-910
2-Fluoro-6-methoxybenzonitrile 95%
2-Fluoro-6-methoxybenzonitrile


Formula: C₈H₆FNO
MW: 151.14
Boiling Pt: 245
Melting Pt: 102…104 °C
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00042291
CAS Number: 94088-46-7
EINECS: 302-047-5
UN: 3439
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty