Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)styrene 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC4018-10G PC4018-1G PC4018-5G
77415-062EA 166.06 USD InStock
77415-062 77415-064 77415-066
3-(Trifluoromethyl)styrene 97%
3-(Trifluoromethyl)styrene


Formula: C₉H₇F₃
Boiling Pt: 64.5 °C/40 mmHg
Density: 1.161 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075481
CAS Number: 402-24-4
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty