Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-5-fluorobenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC4108-100G PC4108-25G PC4108-5G
77415-142EA 247.58 USD InStock
77415-142 77415-144 77415-146
2-Cyano-5-fluorobenzyl bromide
2-Cyano-5-fluorobenzyl bromide


Formula: C₈H₅BrFN
MW: 214.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059542
CAS Number: 421552-12-7
UN: 3265
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty