Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Fluorophenyl)acetonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC4144-100G PC4144-25G
77415-162EA 28.52 USD InStock
77415-162 77415-164
(4-Fluorophenyl)acetonitrile 98%
(4-Fluorophenyl)acetonitrile


Formula: C₈H₆FN
Boiling Pt: 119…120 °C/18 mmHg
Density: 1.126 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001917
CAS Number: 459-22-3
EINECS: 207-286-8
UN: 3276
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty