Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Methyl-4-(trifluoromethyl)benzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC4461-1G PC4461-25G PC4461-5G
77415-428EA 186.02 USD InStock
77415-428 77415-430 77415-432
α-Methyl-4-(trifluoromethyl)benzylamine 98%
alpha-Methyl-4-(trifluoromethyl)benzylamine


Formula: C₉H₁₀F₃N
MW: 189.18 g/mol
Boiling Pt: 76.5…77.5 °C/4.8 mmHg
Density: 1.18 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD04038878
CAS Number: 15996-84-6
UN: 2735
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty