Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-6-methylbenzoic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC53324-1G PC53324-5G
77416-676EA 33.62 USD InStock
77416-676 77416-678
4-Bromo-2-fluoro-6-methylbenzoic acid 95%
4-Bromo-2-fluoro-6-methylbenzoic acid


Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072908
CAS Number: 1242157-23-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty