Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-fluorophenylisocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503001-1G PC503001-25G PC503001-5G
77416-250EA 21.26 USD InStock
77416-250 77416-252 77416-254
2-Bromo-5-fluorophenylisocyanate
2-Bromo-5-fluorophenylisocyanate
May form small amount of sediment.


Formula: C₇H₃BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD09945449
CAS Number: 1016819-26-3
UN: 2206
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty