Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-(trifluoromethyl)phenylacetylene 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC56841-1G PC56841-250MG
77416-906EA 544.04 USD InStock
77416-906 77416-908
2-Bromo-5-(trifluoromethyl)phenylacetylene 95%
2-Bromo-5-(trifluoromethyl)phenylacetylene


Formula: C₉H₄BrF₃
Storage Temperature: Refrigerator
MDL Number: MFCD30529953
CAS Number: 1211523-00-0
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty