Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC5654-1G PC5654-25G PC5654-5G
77416-850EA 21.32 USD InStock
77416-850 77416-852 77416-854
4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride 95%
4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride
Safer alternative to DAST.


Formula: C₁₂H₁₇F₃S
Boiling Pt: 150 °C
Melting Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD11858620
CAS Number: 947725-04-4
UN: 2923
ADR: 8+6.1,I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty