Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluorophenyl isothiocyanate 96%

Supplier: APOLLO SCIENTIFIC
Danger

PC5692-1G PC5692-5G
77416-920EA 124.3 USD InStock
77416-920 77416-922
Pentafluorophenyl isothiocyanate 96%
Pentafluorophenyl isothiocyanate


Formula: C₇F₅NS
Boiling Pt: 109…110 °C/35 mmHg
Density: 1.56 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00041042
CAS Number: 35923-79-6
EINECS: 252-791-9
UN: 2922
ADR: 8+6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty