Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hexafluorosilicate 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC6511-100G PC6511-50G
77417-418EA 24.46 USD InStock
77417-418 77417-420
Sodium hexafluorosilicate 98%
Sodium hexafluorosilicate


Formula: F₆Na₂Si
Density: 2.679 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003491
CAS Number: 16893-85-9
EINECS: 240-934-8
UN: 2674
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty