Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluorothiophenol 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC5820-100G PC5820-25G
77417-118EA 268.83 USD InStock
77417-118 77417-120
Pentafluorothiophenol 99%
Pentafluorothiophenol


Formula: C₆HF₅S
Boiling Pt: 143 °C
Melting Pt: –24 °C
Density: 1.501 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004828
CAS Number: 771-62-0
EINECS: 212-236-3
UN: 2922
ADR: 8+6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty