Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-methylbenzenesulphonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC5941-100G PC5941-25G PC5941-5G
77417-198EA 700.94 USD InStock
77417-198 77417-200 77417-202
2-Fluoro-4-methylbenzenesulphonyl chloride
2-Fluoro-4-methylbenzenesulphonyl chloride


Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
Melting Pt: 42…44 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094330
CAS Number: 518070-29-6
UN: 3265
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty