Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoropyridine-5-carbonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC7055-1G PC7055-250MG
77417-630EA 81.5 USD InStock
77417-630 77417-632
2-Fluoropyridine-5-carbonyl chloride 95%
2-Fluoropyridine-5-carbonyl chloride


Formula: C₆H₃ClFNO
Melting Pt: 275…278 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781233
CAS Number: 65352-94-5
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty