Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobenzyl fluoride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC56794-1G PC56794-250MG PC56794-5G
77416-866EA 206.82 USD InStock
77416-866 77416-868 77416-870
4-Bromobenzyl fluoride 95%
4-Bromobenzyl fluoride


Formula: C₇H₆BrF
MW: 189.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20482167
CAS Number: 459-49-4
UN: 1759
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty