Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Chloro-4-(trifluoromethoxy)toluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7438UF-25G PC7438UF-5G
77417-940EA 24.46 USD InStock
77417-940 77417-942
α-Chloro-4-(trifluoromethoxy)toluene 97%
α-Chloro-4-(trifluoromethoxy)toluene


Formula: C₈H₆ClF₃O
MW: 210.58
Boiling Pt: 72…73 °C/15 mmHg
Density: 1.34 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00052326
CAS Number: 65796-00-1
UN: 3265
ADR: 8 III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty