Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethoxy)phenyl isocyanate 92%

Supplier: APOLLO SCIENTIFIC
Danger

PC7439PW-1G PC7439PW-25G PC7439PW-5G
77417-950EA 20.4 USD InStock
77417-950 77417-952 77417-954
2-(Trifluoromethoxy)phenyl isocyanate 92%
2-(Trifluoromethoxy)phenyl isocyanate
  • May form small amount of sediment.


Formula: C₈H₄F₃NO₂
Boiling Pt: 52…55 °C/15 mmHg
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134160
CAS Number: 182500-26-1
UN: 3080
ADR: 6.1+3,II
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty