Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethynyl-3,5-difluorobenzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC9898-1G PC9898-25G PC9898-5G
77418-752EA 39.74 USD InStock
77418-752 77418-754 77418-756
1-Ethynyl-3,5-difluorobenzene 97%
1-Ethynyl-3,5-difluorobenzene


Formula: C₈H₄F₂
Boiling Pt: 123…124 °C
Density: 1.163 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD04039982
CAS Number: 151361-87-4
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty