Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chloro-5-fluorobenzoyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC6589-1G PC6589-250MG
77417-458EA 197.76 USD InStock
77417-458 77417-460
4-Bromo-2-chloro-5-fluorobenzoyl chloride 95%
4-Bromo-2-chloro-5-fluorobenzoyl chloride


Formula: C₇H₂BrCl₂FO
Storage Temperature: Ambient
MDL Number: MFCD00236590
CAS Number: 203302-93-6
UN: 3265
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty