Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Trifluoromethyl)maleic anhydride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7686-25G PC7686-5G
77418-122EA 358.62 USD InStock
77418-122 77418-124
(Trifluoromethyl)maleic anhydride 97%
(Trifluoromethyl)maleic anhydride


Formula: C₅HF₃O₃
Boiling Pt: 93 °C/65 mmHg
Flash Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153287
CAS Number: 700-27-6
UN: 3265
ADR: 8,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty