Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)thiobenzamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7769F-100G PC7769F-25G PC7769F-5G
77418-178EA 103.98 USD InStock
77418-178 77418-180 77418-182
4-(Trifluoromethyl)thiobenzamide 98%
4-(Trifluoromethyl)thiobenzamide


Formula: C₈H₆F₃NS
Melting Pt: 135…138 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051806
CAS Number: 72505-21-6
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty