Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-fluoropyridine

Supplier: APOLLO SCIENTIFIC
Warning

PC6447-1G PC6447-25G PC6447-5G
77417-392EA 21.32 USD InStock
77417-392 77417-394 77417-396
4-Chloro-3-fluoropyridine
4-Chloro-3-fluoropyridine


Formula: C₅H₃ClFN
MW: 131.54
Boiling Pt: 138…143 °C
Melting Pt: –27…–24 °C
Density: 1.333 g/cm³ (25 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD03453233
CAS Number: 2546-56-7
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty