Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-fluorobenzyl bromide 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC6878-1G PC6878-25G PC6878-5G
77417-584EA 29.54 USD InStock
77417-584 77417-586 77417-588
2-Bromo-3-fluorobenzyl bromide 99%
2-Bromo-3-fluorobenzyl bromide


Formula: C₇H₅Br₂F
MW: 267.923 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12911576
CAS Number: 1184918-22-6
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty