Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)benzenesulphonyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7508-100G PC7508-25G PC7508-5G
77417-990EA 186.44 USD InStock
77417-990 77417-992 77417-994
2-(Trifluoromethyl)benzenesulphonyl chloride 98%
2-(Trifluoromethyl)benzenesulphonyl chloride


Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 133…135 °C/14 mmHg
Density: 1.585 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051696
CAS Number: 776-04-5
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty