Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α-Trifluoro-p-toluonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7562-1KG PC7562-250G
77418-010EA 137.54 USD InStock
77418-010 77418-012
α,α,α-Trifluoro-p-toluonitrile 98%
α,α,α-Trifluoro-p-toluonitrile


Formula: C₈H₄F₃N
Boiling Pt: 183…185 °C
Melting Pt: 37…41 °C
Density: 1.278 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00001826
CAS Number: 455-18-5
EINECS: 207-239-1
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty