Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α'-Chloro-α,α,α-trifluoro-m-xylene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7633F-100G PC7633F-25G
77418-060EA 74.38 USD InStock
77418-060 77418-062
α'-Chloro-α,α,α-trifluoro-m-xylene 98%
α'-Chloro-α,α,α-trifluoro-m-xylene
Derivitising agent for detection of ethyleneurea by electron-capture G.C.


Formula: C₈H₆ClF₃
MW: 194.58
Boiling Pt: 95
Density: 1.254 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00000908
CAS Number: 705-29-3
EINECS: 211-884-4
UN: 2920
ADR: 8+3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty