Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)acrylic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7761-100G PC7761-25G PC7761-5G
77418-166EA 406.5 USD InStock
77418-166 77418-168 77418-170
2-(Trifluoromethyl)acrylic acid 98%
2-(Trifluoromethyl)acrylic acid


Formula: C₄H₃F₃O₂
Boiling Pt: 86 °C/35 mmHg
Melting Pt: 48…52 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00042424
CAS Number: 381-98-6
UN: 3261
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty