Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,4-Trifluoro-1-butanol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7289-1G PC7289-25G PC7289-5G
77417-756EA 24.46 USD InStock
77417-756 77417-758 77417-760
4,4,4-Trifluoro-1-butanol 97%
4,4,4-Trifluoro-1-butanol


Formula: C₄H₇F₃O
Boiling Pt: 122…125 °C
Density: 1.206 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00041483
CAS Number: 461-18-7
UN: 1987
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty