Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,4-Trifluorobutyric acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7292-1G PC7292-25G PC7292-5G
77417-768EA 21.26 USD InStock
77417-768 77417-770 77417-772
4,4,4-Trifluorobutyric acid 98%
4,4,4-Trifluorobutyric acid


Formula: C₄H₅F₃O₂
Boiling Pt: 166…167 °C
Melting Pt: 25…32 °C
Flash Pt: 96.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077604
CAS Number: 406-93-9
UN: 3261
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty