Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3,3-Trifluoro-L-lactic acid

Supplier: APOLLO SCIENTIFIC
Danger

PC8122-1G PC8122-250MG
77418-352EA 730.86 USD InStock
77418-352 77418-354
3,3,3-Trifluoro-L-lactic acid
3,3,3-Trifluoro-L-lactic acid


Formula: C₃H₃F₃O₃
Melting Pt: 66…76 °C
Storage Temperature: Ambient
MDL Number: MFCD04037238
CAS Number: 125995-00-8
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty