Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(dimethylamino)sulfonium difluoro(trimethyl)silicate, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

PC7880-1G PC7880-5G
77418-276EA 159.96 USD InStock
77418-276 77418-278
Tris(dimethylamino)sulfonium difluoro(trimethyl)silicate, Technical Grade
Tris(dimethylamino)sulfonium difluoro(trimethyl)silicate


Formula: C₃H₉F₂Si·C₆H₁₈N₃S
MW: 275.478 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012170
CAS Number: 59218-87-0
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty