Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyldiphenylsilyl trifluoromethanesulfonate

Supplier: APOLLO SCIENTIFIC
Danger

PC7942-1G PC7942-5G
77418-314EA 29.54 USD InStock
77418-314 77418-316
tert-Butyldiphenylsilyl trifluoromethanesulfonate
tert-Butyldiphenylsilyl trifluoromethanesulfonate


Formula: C₁₇H₁₉F₃O₃SSi
Storage Temperature: Refrigerator
MDL Number: MFCD01941212
CAS Number: 92886-86-7
UN: 1760
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty