Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-fluorophenyl isothiocyanate, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

PC9026-25G PC9026-5G
77418-510EA 894.68 USD InStock
77418-510 77418-512
2-Bromo-4-fluorophenyl isothiocyanate, Technical Grade
2-Bromo-4-fluorophenyl isothiocyanate


Formula: C₇H₃BrFNS
Storage Temperature: Ambient
MDL Number: MFCD00673194
CAS Number: 183995-72-4
UN: 2922
ADR: 8+6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty